Logs: A new category is added to the Ardia Server Admin page. This category provides a utility that consolidates logs and metrics, including application metrics, application logs, the system journal, and IQ reports. The utility stores the archived data and allows for easy downloading.