Global Settings - Ardia Core Software - Ardia Core Software 1.1 Release Notes - Ardia Core Software components - Features - Ardia Core Software 1.1 Release Notes - Global Settings - Ardia Core Software
Ardia Core Software 1.1 Release Notes
- Product Tree
- Ardia Platform > Ardia Core Software
- Document Type
- Release Notes
- Software version
- 1.1
- Background verification of raw data is enabled with an option to set the interval between verifications.
- File uploads are restricted to selected extensions only with an option to prevent the upload of executable files and scripts.