Ardia Data Explorer - Ardia Core Software - Ardia Core Software 1.1 Release Notes - Ardia Core Software components - Ardia Data Explorer - Features - Ardia Core Software 1.1 Release Notes - Ardia Core Software
Ardia Core Software 1.1 Release Notes
- Product Tree
- Ardia Platform > Ardia Core Software
- Document Type
- Release Notes
- Software version
- 1.1
- Version management for documents is now available with the following capabilities:
- View version information.
- Compare properties between any two versions, with a summary view that shows differences and a detailed view that lists all properties.
- Revert to any older version.
- The integrity of raw data files can be verified, and any files that fail verification are flagged with a verification failure badge. This feature is configurable in the Ardia Global Settings utility.
- File uploads are restricted to specific file types, which are configured in the Global Settings utility.
- The full audit trail for individual objects in the Ardia Repository can now be opened directly in the Audit Trail Viewer utility as a new browser tab by either:
- Right-clicking the object and selecting View Audit Trail.
- Selecting the object’s row, then selecting View Audit Trail from the header.
- Files such as .pdf, .jpeg, .png, .gif, .tiff, .eps, and .svg can be opened in a new browser tab directly from the contents view.
- Folders, sequences, injections, PDFs, and images can be opened directly from search results.
- Folders and sequences in the content area can be opened by selecting their name.
- Row numbers are introduced for items in the content area.
- A new icon is added to represent the Proteome Discoverer result files.
- The Type column of the content area is updated to display more file types for clarity.
- The Manage Access function, which assigns folder access permissions to specific users and roles, is extended to include the repository-level root folder.
- Multiple items can be selected using SHIFT + select and CTRL + select.
- A drag-and-drop function is implemented to move items to a new location in the content view or in the tree view.
- Audit trail prompts now appear for items that pass validation during the Rename and Move To actions.
- Column customization is now user-specific, meaning that any changes made in a user profile persists even after the user signs off and signs back in.
- The upload progress bar is enhanced to show real-time status of the upload, based on individual file upload progress. Create folders to organize data objects in the Ardia Platform repository.