The actions described in the following table can be performed for a selected user on the Users page.
Function | Subfunction | Description |
---|---|---|
Details | N/A | Displays details for a given user, including the username, identity provider, and account status. Also includes a Resend Invite link, which resends an invite to users whose identity provider is Ardia and whose statuses are as follows: Invite Sent, Pending Login. |
Edit | Edits the identity provider and the account status. If the username is not an email address, the identity provider cannot be changed to Ardia. | |
Edit | N/A | Edits the identity provider and the account status. If the username is not an email address, the identity provider cannot be changed to Ardia. |
Assign | Assign To | Selects roles to be assigned to the user. |
icon (next to selected roles) | Removes the assignment of roles to the user. | |
Duplicate | N/A | Creates a duplicate of the user and allows for modifications to the username, identity provider, and user assignments. The new username must be distinct from those of existing users. |
Disable | Disable User | Changes the status of a user to make it inactive. |
Delete | N/A | Deletes a selected user. |