The Manage and Configure Identity Providers area in the IdP Configuration utility displays all configured IdPs, including the default Ardia IdP. The default Ardia IdP is described in the following table.
Function
Description
Ardia
Indicates the default IdP for the Ardia Platform that comes preconfigured with the Ardia Core Software.
The Manage and Configure Identity Providers area provides the functions described in the following table for users to manage IdPs.
Function
Subfunction
Description
Edit
N/A
Edits a configured IdP.
Add New IdP
N/A
Adds a new IdP by selecting one of the available providers from the Select IdP list.
Select IdP
Contains the following available IdPs:
  • Chromeleon
  • Okta
  • Facebook
  • Google
  • Generic OIDC
  • Thermofisher.com IdP (CDC)
  • Azure AD
  • Twitter
Add
Registers a new IdP by entering the necessary information to configure it.
Remove
Removes any configured IdP (besides the default Ardia Platform IdP) that is no longer needed or is obsolete according to organizational policy.