The Modify Instruments Application Settings area on the Instruments page contains options to define the organizational-specific settings described in the following table for the Ardia Instruments application.
Function | Subfunction | Description |
---|---|---|
Scheduler | N/A | Configures the Ardia Instruments Scheduler global settings. |
What day of the week would you like scheduler to start | Sets Sunday or Monday as the first day of the week for the Scheduler. | |
Require completion of maintenance reservation | Specifies whether the user is required to mark the maintenance as completed in the Scheduler so that the instrument is released for the next reservation. | |
Pick the day when the fiscal year starts | Sets when the fiscal year starts (month and day). |