The Set Platform Settings area on the Platform page contains options to define the organizational-specific administrative settings described in the following table for the Ardia Platform.
Function | Subfunction | Description |
---|---|---|
SSO | N/A | Configures single sign-on (SSO) sessions. |
Enable Timeout Session for Logout | Specifies whether SSO sessions time out. Default state: Off . | |
Timeout Session for Logout | Sets the duration of SSO sessions (in minutes) before a user is automatically logged out due to inactivity. The timer resets if there is any activity. NOTE This subfunction is visible only if the Enable Timeout Sessions for Logout subfunction is enabled . | |
Audit | N/A | Configures audit trail-related settings. |
Prompt for Comments When Saving | Enables or disables the prompt for comments when a user saves any changes. Default state: Off . NOTE If the Prompt for Comments When Saving subfunction is enabled, the Audit Trial Viewer utility records any comments that users enter as user-added comments. | |
Notifications | N/A | Configures global notification settings. |
Retention Time for Cleared Notifications | Specifies the number of days before the application clears all notifications. | |
Users | N/A | Configures user settings that affect all users. |
Delete Users | Allows users to be deleted by users with appropriate administrative privileges. Default state: Off . | |
Disable and Reactivate Users | Allows users to be disabled or reactivated by users with appropriate administrative privileges. Default state: Off . |