The Client Registration and Management utility lists the previously generated registration codes and their details. The parameters described in the following table are used to define the codes.
Parameter | Description |
---|---|
App Name | Displays the name of the application that is connected to the Ardia Platform, such as BioPharma Finder or the Chromeleon CDS. |
Version | Displays the release version of the registered application. |
Status | Indicates the status of registration codes as follows:
|
Reg Code | Displays the eight-digit alphanumeric code used by the application to connect to the Ardia Platform. |
Code Date | Displays the date when the registration code was created. |
Code Age | Displays the number of days since the registration code was last used. |
PC Name | Displays the name of the server or client that was used to connect to the Ardia Platform to access an application or application environment. |
IPv4 Address | Displays the IP address from which the connection to the application originated. |