The Client Registration and Management utility allows users to manage client application registrations using the actions described in the following table.
Function | Subfunction | Description |
---|---|---|
Actions | Add Registration Code | Generates a new registration code that is required for a client application to register with the Ardia Platform. Displays the platform URL and code in a dialog box for easy copying and pasting. |
Copy Code | Redisplays the platform URL and code in a dialog box for copying and pasting when needed. | |
Disable Apps | Temporarily prevents a client application from connecting to the platform. | |
Enable Codes | Restores a disabled registration code. | |
Remove Apps | Permanently deregisters a client application from the platform. After deregistration, the application can no longer connect to the platform and is removed from the list of registered applications. |