The navigation pane displays the Ardia Repository folders in a hierarchical tree structure. A context menu appears when right-clicking a folder in the tree view. Users can perform the actions described in the following table from this context menu.
Function | Description |
---|---|
Pin to Quick Access | Adds a selected item to the Quick Access list. |
Remove from Quick Access | Removes a selected item from the Quick Access list. |
Delete | Deletes a selected item from the tree view. |