The functions described in the following table are available for running, loading, and saving queries.
Function | Description |
---|---|
Run Query | Executes the current query conditions and displays results in the audit entry table. |
Load Query | Opens a dialog box to select a previously saved query. The conditions from the selected query are added to the query builder. |
Save Query | Opens a dialog box to select a location and name for the query. The current conditions that are displayed in the query builder are saved. |