Audit entries are displayed in a table format with the parameters described in the following table.
Function | Description |
---|---|
Timestamp | Specifies the time at which an audited event occurs. It is displayed in the local time and date format of the browser location. In the US, it is displayed as mm/dd/yyyy, hh:mm:ss AM/PM, and in Europe, it is displayed as dd/mm/yyyy, hh:mm:ss. The tooltip displays the equivalent time in Coordinated Universal Time (UTC) format (yyyy-mm-ddThh:mm:ssZ), where T stands for time and separates the date and time components, while Z stands for Zulu time and indicates that the time is in UTC time zone. |
Object Type | Specifies the type of object that triggers the audit record. |
Object Name | Specifies the name of the object that triggers the audit record. |
Application | Specifies the name of the application or service in which the audit event occurred. |
Action | Specifies the type of activity that is performed and recorded in the audit log. |
Username | Specifies the name of the user who performs the audited event. |
Role | Specifies the role that the user is assigned to at the time the audited event is performed. |
Comment | Specifies user-added comments (if the user has provided a comment). |
Path | Specifies the Ardia Repository file path of the object that triggers the audit record. |
Additional Info | Captures any system-generated additional information, if available. |