The Audit Trail Viewer provides a dropdown list to filter audit entries by type. The available types are described in the following table.
Function | Description |
---|---|
All (selected by default) | Displays events from all types of audit trails. |
Authentication | Displays logon and logoff events. |
Data Applications | Displays events that are related to data, including events from the Data Explorer application and connected applications. |
Global Settings | Displays events that are related to the Global Settings utility. |
External Connections | Displays events that are related to connecting applications or identity providers (IdPs) to the Ardia Platform. |
User Management | Displays events that are related to user management objects. |