The Audit Trail Viewer utility displays entries that are generated automatically on the Ardia Platform when users perform specific actions.
This utility allows users to create queries to filter entries based on specific types and save them for future use.
The Audit Trail Viewer utility consists of the following four main areas:
  • Audit Trail Type list: Allows users to filter entries by type.
  • Audit entry table: Displays entries with specific information about the action or actions being audited.
  • Queries: Allows users to limit entries using user-defined rules.
  • Page navigation: Allows users to navigate through a large number of audit entries.