The Data Explorer application has some functions that are always available, regardless of the item selection. These functions are located above the content area and are described in the following table.
Function
Subfunction
Description
Search
N/A
Allows users to quickly locate an item in the current folder.
New Folder
N/A
Creates a new folder inside the current folder.
Upload Files
N/A
Allows users to upload files to the Ardia Platform repository. Depending on the selected item and destination container types, the Upload Files function works differently.
Upload any item other than a raw data file to a folder
Uploads the selected items to the current folder.
Upload a raw data file to a sequence
Uploads raw data files to the current sequence.
Upload a raw data file to a folder
Creates a new sequence with the selected raw data files.
Cancel an Upload
Cancel the upload of an item
Cancels the uploading of a specific item.
Cancel all uploads
Cancels all the uploading processes.