The content area displays the contents of the folder selected in the navigation pane. The contents are displayed in a table format, where each row corresponds to an item and the columns represent associated parameters. The following table describes the parameters in the content area.
Parameter | Applicable item type | Description |
---|---|---|
Name | All | Specifies the name of the item. |
Type | All | (For injections) Specifies the injection type. Examples: Calibration Standard, Check Standard, Blank, Matrix, Spiked, Unspiked, and Unknown. (For items other than injections) Specifies the category of the item. Examples: Folder, Sequence, Audit Query, Peptide Experiment, and so on. |
Size | Except Folders and Sequences | Specifies the size of the item. |
Date Modified | All | Specifies the date and time when the item was last modified. |
Modified By | All | Specifies the user who last modified the item. |
Status | All | Specifies the current status of the item. The status of an item can be one of the following:
|
Position | Injection | Specifies the position of the injection. |
Acquisition Status | Injection | Specifies the current status of the injection acquisition. The status can be one of the following:
|
Inject Time | Injection | Specifies the date and time at which the injection occurred. |
Upload Status | Injection | Specifies the current status of the uploading process of the injection. The status can be one of the following:
|
Result Path | Injection | Specifies the folder path of the location where the result of the injection is saved. |
Edit Columns | N/A | Allows users to do the following:
|