The root folder in the Ardia Platform repository is named Ardia Repository. By default, every Data Explorer user is granted the Full Access privilege to this folder.
The root folder in the Ardia Platform repository is named Ardia Repository. By default, every Data Explorer user is granted the Full Access privilege to this folder.