This guide outlines the basic steps to connect your instruments to the Ardia Instruments application in the Thermo Scientific™ Ardia™ Platform. With the Instruments application, you can remotely manage, schedule, and monitor data acquisition on your instruments.
Before you start
The Ardia Platform Link application must be installed and set up on each instrument PC.
Steps
1. Sign in to the Ardia platform
Open the Ardia platform URL in a browser and sign in to the Ardia platform.
2. Launch the Ardia Instruments application
Open the Instruments application from the Ardia platform Home page
3. (Optional) Connect your lab to Thermo Fisher
Select Continue to connect. If you do not have permission, use the Settings page to connect later.
4. Set up your system
On the Waiting for Setup page, select Complete Setup on the card of your instrument.
5. Finish setup
Follow the on-screen instructions to enter the required instrument information, and then select Finish Setup.
6. Start managing connected systems
On the Active page, view all your newly set up systems and manage your lab in one place.
© 2023 Thermo Fisher Scientific Inc. All rights reserved.
All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
Pub. No. BRE0062367, Revision A
For Research Use Only. Not for use in diagnostic procedures.