The movement of the robot arm is controlled as a safety measure as follows:
- During the startup and initialization of the instrument, the robot arm moves at a slower than usual speed.
- If a collision occurs involving the robot arm, the motor of the robot arm shuts down.