Before you begin to load the AccelerOme instrument deck:
- Ensure the instrument installation and setup procedures are completed.
- Ensure the IQ test is executed successfully.
- Ensure the instrument is ready for use.
Load the instrument deck as follows.
While loading the instrument deck:
- Leave the vial positions not needed for the intended procedures empty.
- Do not use the tray positions C, E, and F on the deck for storing vials, reagents, or flammable liquids or for any other purpose. They are meant to be unused.
NOTE
Remove the bottle caps of the peptide clean-up buffers (tray D buffers) before use.