The navigation paths to a certain command or item are displayed according to the following convention.
Path (in User level or Extended User level): Start Screen -> Options -> Maintenance -> Create Configuration Backup |
Each path starts from the Start Screen, which is the home screen of the PAL firmware on the PAL terminal. The preceding line indicates if the path is followed in theUser level or the Extended User level.
TIP
Pressing the back button continuously brings you back to the start screen.