Follow the subsequent considerations for waste fluid disposal:
- Collect the waste fluid into a container suitable for the respective type of waste according to the applicable national, regional or local regulations for bio, radio or chemical hazardous substances.
- Handle the waste fluid and dispose it according to the applicable national, regional or local regulations for bio, radio or chemical hazardous substances.
- Verify that the waste line is correctly and tightly connected, by purging it with clean water. Visually check if water is leaking out of any of the connections and reconnect the connections, as necessary.
- If the waste line is pinched, the waste fluid flow is blocked and this leads to an interruption of the work flow.
- Verify that the waste line is routed correctly without any pinching, by visually checking the entire routing.
CAUTION
Biological Hazard
If the waste line is torn off, the waste fluid can contaminate your working area and causing health impairing injury.