Consumable | Description |
|---|---|
Dilutor tool syringe | The dilutor syringe is regarded as a consumable due to the wear down of its plunger and seal |
Dilutor needle | The dilutor needle is regarded as a consumable due to the wear down and/or damage or bending |
Sample preparation fluids | Permitted sample preparation fluids are only the ones provided by Thermo Scientific™ |
Buffer fluids | Permitted buffer fluids are only the ones provided by Thermo Scientific™ |