The instrument ships with the following tray inserts and solution tray adapters:
  • Output plate tray insert (labeled Insert ABGene plate)
  • Reagent kits tray insert (labeled Insert reagent vials)
  • Reagent kits solution tray adapter (labeled B1/B2)
  • D buffer tray adapter (labeled D)
Steps
  1. Install the output plate tray insert (labeled Insert ABGene plate) into the tray slot labeled A.
  2. Output plate insert
    Output plate insert
  3. Install the reagent kits tray insert (labeled Insert reagent vials) into the tray slot labeled B1/B2.
  4. Reagent kits insert
    Reagent kits insert
  5. Install the reagent kits adapter (labeled B1/B2) on top of this insert.
  6. Reagent kits adapter
    Reagent kits adapter
  7. Install the buffer tray adapter plate (labeled D) into the tray slot labeled D.
D buffer tray adapter