The Install Qualification (IQ) kit consists of an output plate and two vials of the protein digestion reagent kit (blue caps) in position 4 and 5 of the kit. This kit is used to run an install qualification test after installing all the consumables in the AccelerOme instrument, to verify if the installation procedure was performed accurately.
Install the protein digestion reagent kit (blue caps) with two vials in position 4 and 5, in the front part of the reagent kits solution tray adapter.
IQ kit
IQ kit