If the instrument front door is open, the power on self test does not start when you turn the instrument on and the instrument interface displays, Power on test cannot start because door is open.
To troubleshoot this error, close the front door and run the system test from the service tools menu on the instrument interface to check if the issue is resolved.