The needle wash solutions are not included with the instrument. Prepare one liter of each of the following solutions:
AccelerOme system needle wash solutions composition
Solution name
Solution composition
Needle Wash Solution W1
Needle Wash Solution W1 – 0.1% formic acid in water, LC-MS grade
(for example, Water with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade)
Needle Wash Solution W2
Needle Wash Solution W2 – 50% methanol and 0.05% formic acid in water, LC-MS grade
(for example, 1:1 v/v mix of needle wash solution W1 and Optima™ LC/MS Grade methanol)