Follow the preventative maintenance schedule for periodically checking the AccelerOme instrument modules for damage or wear and replace them as and when necessary.
Maintenance procedure | Frequency |
|---|---|
Clean the instrument | Monthly |
Check and replace the solution tubes | Monthly |
Check and replace the waste line | Monthly |
Check teaching and perform install qualification (IQ) test | Annually |
Check the dilutor syringe for any damage or wearing and replace if necessary. | Annually |
Check if the flow path of the syringe is clear. | Annually |
Check for leaks at the syringe/valve connection. | Annually |
Check if the liquid drains to the waste container upon priming the solution lines. | Annually |