Your laboratory must have one or more RJ-45 wall outlets to connect the AccelerOme instrument to the network.
You are responsible for connecting the instrument to your network.
Equipment | Heat Output (BTU per Hr.) | Heat Output (in W) |
---|---|---|
AccelerOme | 717 | 210 |