Parameter | Requirements |
|---|---|
Protection Class | Class I |
Wet location | No |
Over voltage category | Category II |
Pollution degree | 2 |
Moisture protection | Normal (IPX0) |
Voltage | 100-240 VAC 50/60 Hz |
Fuse (PAL control board) Fuse (power inlet socket) | T6.3A/250 V T4A/250 V (2 pcs) |
Ensure your site has enough power to support the system.
Equipment | Maximum Power (VA) |
|---|---|
AccelerOme | 250 VA |
Ensure to have a power outlet with a protective contact/ground contact that supplies the operating voltage required for the device. Consult a qualified electrician to install this outlet.