Ensure to maintain good electrical grounding for the power line. Poor grounding is dangerous and can severely affect the instrument performance.
Do not connect the AccelerOme instrument to power lines used for heavy-duty devices, such as motors, UV lamps, refrigerators, air compressors and other heavy-duty devices that can generate disturbances.
Low quality power lines can adversely affect the system operation. You are responsible to correct the line voltage problems. Power conditioning equipment must be specified by a company or a consultant specialized in that field. Contact your Thermo Fisher Scientific Field Service Engineer (FSE) for assistance in locating a power consultant.