To ensure the entrance to your site accommodates the AccelerOme instrument:
- Ensure the width of your door opening is at least 80 cm (32 in).
- Make sure you have enough room to move the delivery box around corners, into elevators, and through doorways.
The dimensions and weight of the shipping box containing the AccelerOme instrument are stated as follows:
Container | Depth (Y-axis) | Width (X-axis) | Height (Z-axis) | Mass | ||||
|---|---|---|---|---|---|---|---|---|
cm | in | cm | in | cm | in | kg | lbs | |
AccelerOme | 97 | 38 | 77 | 30 | 110 | 43 | 75 | 165 |