The instrument front, left side, and back design is described as follows:
| No. | Description | No. | Description |
|---|---|---|---|
1 | Front door to the instrument compartment | 3 | Instrument interface to control the instrument software |
2 | Two front USB ports (for example, to connect the barcode scanner) | 4 | Door to the service compartment with the PAL terminal and the power switch |
| No. | Description | No. | Description |
|---|---|---|---|
1 | Two fans for cooling and exhaust air | 2 | Sockets for the power cord and ethernet cable. The socket for the power cord has an integrated fuse. |