Statistical power is the likelihood of the analytical approach to detect a significant change when the biology in the study changes.
Click View Details to see the detailed statistical power estimate information.
Statistical power estimate
Statistical power estimate
The estimate is made apriori for each study based on:
  • Statistical model assumptions of a two group two tailed T-test with a fixed significance cutoff p=0.05.
  • Adjustable values of expected analyte coefficient of variation and minimum biological effect.
  • When more than two groups have been defined, pairwise comparisons can be assumed. The group having the smallest n is used for the apriori estimate calculation.