Parameter | Description |
|---|---|
Prime/Empty Lines | Primes or empties the solution lines. |
Back | Returns to previous screen. |
Prime | Runs the priming and emptying script. |
Cancel | Stops the priming and emptying script. |
System Test | Homes the robot arm, performs a spectrophotometer test and a thermal mixer quick test. |
Run Test | Starts the system test. |
Back | Returns to previous screen. |
Cancel | Cancels the test. |
Thermomixer Test | Performs a thermomixer ramp test. |
Back | Returns to previous screen. |
Run Test | Starts the thermomixer temperature ramp test. |
Cancel | Cancels the test. |
Change the Needle | Changes the needle on the dilutor tool. |
Back | Returns to previous screen. |
Next | Advances to the next screen. |
Download "How to" PDF to USB | Saves the instructions on a USB drive (if available). |
Done | Returns to the Service Tools screen. |
Event Log | View the event log of the system. |
Filter On | Shows events matching the criteria: Run History, Service, Settings, System, Audit, Alarm, Error, Alert, or Method. |
Export | Exports the specified events with options for further specification. |
Format | Specifies the format for the event report (CSV or PDF). |
Events to Export | Select events to export: Run History, Service, Settings, System, Audit, Alarm, Error, Alert, or Method. |
Date Range | Specifies the date range: 1 day, 7, days, 60 days, or a custom range. |
USB drive | Sends exported report file to the USB drive in the instrument. |
Network Drive | Sends exported report file to the desktop. |
Additional Export | Returns to the Event Log screen. |
Done | Returns to the home screen. |
Tray Cooler | Sets the tray cooler temperature. |
On/Off | Turns the tray cooler on or off. |
Temperature selector | Select the tray cooler temperature. |
Save | Saves the selected temperature. |
Qualification Test | Runs the install qualification test on the instrument. |
Back | Returns to previous screen. |
Run Test | Starts running the install qualification test. |
Next | Moves to the next step in guided setup. |
Start | Starts running the install qualification test after the completion of guided setup. |
Cancel | Cancels the install qualification test run or the deck validation run. |
Unload Instrument | Provides instructions to unload the instrument. |
View Results | Provides test result information and allows exporting this data. |
Done | Returns to the home screen. |
Re-measure Qualification Samples | Perform a re-measurement of qualification samples. |
Run Test | Starts running the re-measurement of qualification samples. |
Back | Returns to previous screen. |
Next | Moves to the next step in guided setup. |
Start | Starts running the install qualification test after the completion of guided setup. |
Cancel | Cancels the test. |
Unload Instrument | Provides instructions to unload the instrument. |
Done | Returns to the home screen. |
Qualification Test History | Displays a list of the results of qualification tests run on the instrument. |
Export | Exports the qualification test history. |
Delete | Deletes the qualification test results. |
Shut Down Instrument | Shuts down the instrument. |
No | Does not shut down the instrument. |
Yes | Shuts down the instrument. |