Shut down the instrument as follows.
Steps
  1. On the home screen, tap the Settings icon (gear), tap Service Tools, and then tap Shut Down Instrument.
  2. Tap Yes to shut down the instrument. The robot arm moves to the tool change station and the instrument interface screen turns black.
  3. Open the service compartment and switch off the power switch to completely turn the instrument off.
Manual shut down
If the shut down function from the instrument interface does not work or it cannot be accessed, manually turn the instrument off using the power switch in the service compartment and then manually move the robot arm down into the tool change station to prevent damage to the needle.
Move robot arm into tool change station
Move robot arm into tool change station
Robot arm resting in tool change station