This service tool executes the measurement part of the installation qualification test, so that if there is an issue with the test results, the user can re-measure the samples after troubleshooting the issue.
Steps
- On the home screen, tap the Settings icon (gear), tap Service Tools, and then tap Re-measure Qualification Samples.
- Ensure the system is ready and then tap Run Test.
- Move through the guided setup steps (as stated below) by tapping Next and at the end of the guided setup, tap Start.
- Ensure that each needle wash bottle (W1 and W2) contains at least 250 mL of each solution.
- Load the output plate in position A.
NOTE
Clear any alarms and alerts after the run by resolving and acknowledging them.