An install qualification test verifies if all the consumables are properly installed in the instrument.
Prime the lines before running an install qualification test.
Steps
- On the home screen, tap the Settings icon (gear), tap Service Tools, and then tap Qualification Test.
- Ensure the system is ready to run the installation test and then tap Run Test.
- Move through the guided setup steps (stated below) by tapping Next and at the end of the guided setup, tap Start.
- Ensure that each needle wash bottle (W1 and W2) contains at least 250 mL of each solution.
- Load the reagent module in position B1.
- Load the output plate in position A.
NOTE
Clear any alarms and alerts after the run by resolving and acknowledging them.