The event log contains information regarding the run history on the instrument as well as the error logs.
Steps
- On the home screen, tap the Settings icon (gear), tap Service Tools, and then tap Event Log.
- (Optional) On the Event Log screen, tap Filter On from the View All drop-down menu and select the types of events to show in the filtered events.
- (Optional) Expand the widget on the events to view further information.
- Tap Export.
- On the Export Event Log screen:
- Select a format for the exported file: CSV or PDF.
- Tap Filter On and change the filtered events.
- Select a date range.