Perform tool calibration in the Extended User level.
For the PAL terminal button functions, refer to the PAL terminal section.
Steps
- On the PAL terminal screen, select DIL UV 1; NL: 80 mm.
- Select Options and select Calibrate Tool. Select Magn2mL.
- Move the robot arm manually to the teach point position displayed on the PAL terminal screen. Lower the tool so it touches the teach point and align it so that the tool is positioned at the center of the teach point.
- Press Save on the PAL terminal. This displays the coordinates of the saved position and allows you to manually adjust the coordinate values, if necessary. To change the coordinate values for either the x, y, or z position, select the respective co-ordinate and edit the value using the scroll wheel.
- When done, press Next. Press OK.