The base of the instrument interior on which all the consumables and kits are loaded is called the instrument interior deck.
The AccelerOme instrument interior deck with and without the tray cooler installed are shown below.
AccelerOme deck with tray cooler
1234567891011
No.DescriptionNo.Description
1
Electrical connections
7
Thermal mixer
2
Service area
8
Solution compartment
3
Tray cooler
9
Tray holder 1
4
Reference point
10
PAL terminal
5
USB connections
11
Power switch
6
Instrument interface
AccelerOme deck with tray holder 2
1234567891011
No.DescriptionNo.Description
1
Electrical connections
7
Thermal mixer
2
Service area
8
Solution compartment
3
Tray holder 2
9
Tray holder 1
4
Reference point
10
PAL terminal
5
USB connections
11
Power switch
6
Instrument interface
CAUTION
Hand Injury
Do not reach into the instrument interior deck (beyond the yellow safety strip) when the instrument is running and the robot arm is moving.
Instrument interior deck safety strip
Instrument interior deck safety strip