Check the solution tubes monthly for any damage or wear. Replace them, if necessary.
Steps
- Visually check the solution tubes in the instrument interior and in the solution compartment for deposits of dirt, flexing, damage, or leaks.
- Replace the solution tubes that are contaminated with dirt deposits, are flexed, damaged, or leaking.
TIP
Check and refill the needle wash solutions monthly.