Change the reconstitution parameters for the injection amount and volume as follows.
Steps
- On the home screen, tap the Settings icon (gear) and tap Reconstitution for LCMS.
- Change one or both of the following settings:
- Select a new value for LCMS Injection Amount.
- Select a new value for LCMS Injection Volume.
Reconstitution parameters
Parameter | Description |
|---|---|
LCMS Injection Amount | Sets the LCMS injection amount. The boundary conditions are 0.01 micrograms to 1 micrograms in 0.01 microgram increments between 0.01 and 0.3, then 0.1 increments. Default = 0.2 micrograms. |
LCMS Injection Volume | Sets the LCMS injection volume. The boundary conditions are 0.1 to 6 microliters in 0.05 microliter increments. Default = 1 microliters. |
Save | Saves changes. Only active when a change has been made. |