When the sample preparation run is complete, the output tray has clean peptides in elution solution. In order to inject for standard (reverse phase chromatography) LCMS analysis, the samples must be dried and re-suspended in sample reconstitution solution for LCMS injection.
For more details, refer to the reagent kit instructions.