The barcode scanner is used to scan the reagent and solution kits to run an experiment. To install the barcode scanner, plug the USB cable of the scanner into the USB port on the front of the instrument and plug the connector on the other end into the barcode scanner.