The dilutor syringe sucks in the needle wash solutions from the needle wash solution bottles in the solution compartment and the dilutor tool transfers them to the needle.
Steps
  1. Unscrew the screw that affixes the syringe to the dilutor module.
  2. Unscrew the syringe screw
    Unscrew the syringe screw
  3. Install the syringe into the provided slot in the 6-port valve.
  4. Install syringe
    Install syringe
  5. Reattach the screw to its original location affixing the syringe to the module.
Reattach syringe screw
Reattach syringe screw