This section describes the installation process for the Install Qualification kit and the solution tray adapters.
NOTE
Install the protein digestion reagent kit (blue caps) with two vials in position 4 and 5 only before running the IQ test.