A flat wrench is required to level the instrument by adjusting the height of the instrument feet. This flat wrench is available in the service compartment on the left side of the instrument.
A flat wrench is required to level the instrument by adjusting the height of the instrument feet. This flat wrench is available in the service compartment on the left side of the instrument.