Perform check teaching in the User level.
For the PAL terminal button functions, refer to the PAL terminal section.
Steps
  1. On the PAL terminal screen, select Tray Cooler 1. Select Slot 2. Select Tray Adp 1. Select B1 slot, B1_Reagents, and then VT16 Rgt.
  2. Select tray cooler 1
    Select tray cooler 1
    Select slot 2
    Select slot 2
    Select tray adp1
    Select tray adp1
    Select B1 slot
    Select B1 slot
    Select B1 reagents
    Select B1 reagents
  3. Select Options and select Check Teaching.
  4. Select check teaching
    Select check teaching
  5. The on screen graphic displays the teaching points for tray B1 (There are three teach points for each: back left, front left, and front right edge positions of the module). Before checking teaching, ensure that three kit vials are placed in the three teach point positions. For checking the first teach point, press Check. The robot arm moves to the teach point and checks teaching by lowering the tool to the back left edge position of tray B1 (so it touches the vial cap) and verifying the correct position and coordinates along the X, Y, and Z axes. When positioned correctly, the tool aligns with the center of the teach point position.
  6. Check first teach point
    Check first teach point
    Tray B1 first teach point
    Tray B1 first teach point
  7. When done, press Next.
  8. Press next
    Press next
  9. Press Check to check the teaching on the second teach point at the front left edge of tray B1.
  10. Check second teach point
    Check second teach point
    Tray B1 second teach point
    Tray B1 second teach point
    Press next
    Press next
  11. Repeat this for the third teach point at the front right. When the teaching is checked for tray B1, press Next on the PAL terminal.
  12. Check third teach point
    Check third teach point
    Tray B1 third teach point
    Tray B1 third teach point
    Press next
    Press next
  13. Press OK. This moves the robot arm back to the home position.
Check teaching complete
Check teaching complete
NOTE
If the check teaching fails (for example, the position of the module is incorrectly aligned with respect to the tool), perform a re-teaching of the module (see Tray B1 teaching).