Perform check teaching in the User level.
For the PAL terminal button functions,refer to the PAL terminal section.
Steps
- On the PAL terminal screen, select Tray Cooler 1. Select Slot 1.
- Select Options and select Check Teaching.
- The on-screen graphic displays the teaching point for the tray cooler, which is located at the bottom center of the B1/B2 tray slot. For checking the teach point, remove the kits, B1/B2 trays, the tray adapter, and the tray insert. Press Check. The robot arm moves to the teach point and checks teaching by lowering the tool to the teach point and verifying the correct position and coordinates along the X, Y, and Z axes. When positioned correctly, the tool aligns with the center of the teach point position.
- When done, press Next.
- Press OK. This moves the robot arm back to the home position.
NOTE
If the check teaching fails (for example, the position of the module is incorrectly aligned with respect to the tool), perform a re-teaching of the module (see Tray cooler teaching).